Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Iodo-3-(trifluoromethoxy)benzene, 97+%

Click to view available options
Quantity:
1g
5g
198206-33-6
C7H4F3IO
288.008
MFCD01090992
UQZXQSQWKJZHCD-UHFFFAOYSA-N
1-iodo-3-trifluoromethoxy benzene,3-trifluoromethoxy iodobenzene,3-iodo-1-trifluoromethoxy benzene,m-iodotrifluoromethoxybenzene,1-iodo-3-trifluoromethoxy-benzene,benzene, 1-iodo-3-trifluoromethoxy,pubchem4438,acmc-1c5ss,3-trifluoromethoxyiodobenzene,ksc494i7t
2777293
1-iodo-3-(trifluoromethoxy)benzene
C1=CC(=CC(=C1)I)OC(F)(F)F
198206-33-6
C7H4F3IO
288.008
MFCD01090992
UQZXQSQWKJZHCD-UHFFFAOYSA-N
1-iodo-3-trifluoromethoxy benzene,3-trifluoromethoxy iodobenzene,3-iodo-1-trifluoromethoxy benzene,m-iodotrifluoromethoxybenzene,1-iodo-3-trifluoromethoxy-benzene,benzene, 1-iodo-3-trifluoromethoxy,pubchem4438,acmc-1c5ss,3-trifluoromethoxyiodobenzene,ksc494i7t
2777293
1-iodo-3-(trifluoromethoxy)benzene
C1=CC(=CC(=C1)I)OC(F)(F)F
Quantity 5g
Flash Point 65°C (150°F)
Boiling Point 185°C to 186°C
Refractive Index 1.52
Sensitivity Light sensitive
Density 1.855
Formula Weight 288.01
Percent Purity ≥97%
Chemical Name or Material 1-Iodo-3-(trifluoromethoxy)benzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures