Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 1-Iodo-4-isopropylbenzene, 97%
Click to view available options
Quantity:
5g
25g
Specifications
Refractive Index | 1.578 |
Sensitivity | Light sensitive |
Density | 1.525 |
Quantity | 5g |
Boiling Point | 236°C to 238°C |
Beilstein | 2205506 |
Formula Weight | 246.09 |
Percent Purity | 97% |
Chemical Name or Material | 1-Iodo-4-isopropylbenzene |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 241-384-1
TSCA : No
Recommended Storage : Ambient temperatures