Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Iodo-4-nitrobenzene, 98+%

Click to view available options
Quantity:
5g
25g
100g
636-98-6
C6H4INO2
249.007
MFCD00007299
SCCCFNJTCDSLCY-UHFFFAOYSA-N
benzene, 1-iodo-4-nitro,p-iodonitrobenzene,4-iodonitrobenzene,p-nitroiodobenzene,4-nitroiodobenzene,4-iodo-nitrobenzene,p-nitrophenyl iodide,1-iodo-4-nitro-benzene,4-iodo-1-nitrobenzene,4-nitro-1-iodobenzene
12495
1-iodo-4-nitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])I
636-98-6
C6H4INO2
249.007
MFCD00007299
SCCCFNJTCDSLCY-UHFFFAOYSA-N
benzene, 1-iodo-4-nitro,p-iodonitrobenzene,4-iodonitrobenzene,p-nitroiodobenzene,4-nitroiodobenzene,4-iodo-nitrobenzene,p-nitrophenyl iodide,1-iodo-4-nitro-benzene,4-iodo-1-nitrobenzene,4-nitro-1-iodobenzene
12495
1-iodo-4-nitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])I
Applications
1-Iodo-4-nitrobenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
Boiling Point 288°C to 290°C
Quantity 25g
Melting Point 172°C to 177°C
Beilstein 1100378
Formula Weight 249.01
Percent Purity ≥98%
Chemical Name or Material 1-Iodo-4-nitrobenzene

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA