Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Iodo-4-(trifluoromethoxy)benzene, 98%

Click to view available options
Quantity:
1g
5g
25g
103962-05-6
C7H4F3IO
288.008
MFCD00042411
RTUDBROGOZBBIC-UHFFFAOYSA-N
1-iodo-4-trifluoromethoxy benzene,4-trifluoromethoxy iodobenzene,4-iodotrifluoromethoxybenzene,1-iado-4-trifluoromethoxy benzene,p-trifluoromethoxy iodobenzene,4-trifluoromethoxy-iodobenzene,1-iodo-4-trifluoromethoxy-benzene,benzene, 1-iodo-4-trifluoromethoxy,alpha,alpha,alpha-trifluoro-4-iodoanisole,4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene
2777294
1-iodo-4-(trifluoromethoxy)benzene
C1=CC(=CC=C1OC(F)(F)F)I
103962-05-6
C7H4F3IO
288.008
MFCD00042411
RTUDBROGOZBBIC-UHFFFAOYSA-N
1-iodo-4-trifluoromethoxy benzene,4-trifluoromethoxy iodobenzene,4-iodotrifluoromethoxybenzene,1-iado-4-trifluoromethoxy benzene,p-trifluoromethoxy iodobenzene,4-trifluoromethoxy-iodobenzene,1-iodo-4-trifluoromethoxy-benzene,benzene, 1-iodo-4-trifluoromethoxy,alpha,alpha,alpha-trifluoro-4-iodoanisole,4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene
2777294
1-iodo-4-(trifluoromethoxy)benzene
C1=CC(=CC=C1OC(F)(F)F)I
Flash Point 77°C (170°F)
Quantity 25g
Boiling Point 72°C to 74°C (15mmHg)
Beilstein 6193794
Formula Weight 288.01
Percent Purity 98%
Chemical Name or Material 1-Iodo-4-(trifluoromethoxy)benzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning