Alfa Aesar™ 1-Iodododecane, 98%, stab. with copper

Manufacturer: Alfa Aesar™ A1190936

Quantity 500g

 View more versions of this product

Catalog No. AAA1190936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4292-19-7
    Molecular Formula CH3(CH2)11I
    Formula Weight 296.24
    MDL Number MFCD00001088
    Beilstein 1738943
    Synonym n-Dodecyl iodide; Lauryl iodide
    Chemical Name or Material 1-Iodododecane, Stabilized with copper

    Specifications

    Formula Weight 296.24
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 128° to 130°C (11mmHg)
    Melting Point −3°C
    Beilstein 1738943
    Synonym n-Dodecyl iodide; Lauryl iodide
    Health Hazard 1 Warning
    MDL Number MFCD00001088
    CAS 4292-19-7
    Quantity 500g
    TSCA TSCA
    Molecular Formula CH3(CH2)11I
    Flash Point >110°C (230°F)
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 1-Iodododecane, Stabilized with copper
    Assay 98%

    Description

    Applications
    1-Iodododecane has been used in the preparation of dodecylated graphite, alkyl functionalized single walled carbon nanotubes (SWNTs). It is also used as chemical intermediate.

    Solubility
    Not miscible or difficult to mix in water.

    Notes
    Light Sensitive. Keep away from oxidizing agents. Store in dark.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit