Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Iodoheptane, 98%, stab. with copper

Click to view available options
Quantity:
50g
250g
4282-40-0
C7H15I
226.101
MFCD00001104
LMHCYRULPLGEEZ-UHFFFAOYSA-N
heptyl iodide,n-heptyl iodide,heptane, 1-iodo,iodoheptane,n-heptyliodide,iodo heptane,1-iodanylheptane,1-iodomeptane,1-iodo heptane,1-iodo-heptane
20274
1-iodoheptane
CCCCCCCI
4282-40-0
C7H15I
226.101
MFCD00001104
LMHCYRULPLGEEZ-UHFFFAOYSA-N
heptyl iodide,n-heptyl iodide,heptane, 1-iodo,iodoheptane,n-heptyliodide,iodo heptane,1-iodanylheptane,1-iodomeptane,1-iodo heptane,1-iodo-heptane
20274
1-iodoheptane
CCCCCCCI
Applications
1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
Melting Point -48°C
Flash Point 78°C (174°F)
Quantity 250g
Boiling Point 203°C to 205°C
Formula Weight 226.1
Percent Purity 98%
Chemical Name or Material 1-Iodoheptane, stabilized with copper

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MJ1050000

TSCA : TSCA