Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodohexane, 98%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3758236
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
638-45-9
C6H13I
212.07
MFCD00001102
ANOOTOPTCJRUPK-UHFFFAOYSA-N
12527
1-iodohexane
CCCCCCI
638-45-9
C6H13I
212.07
MFCD00001102
ANOOTOPTCJRUPK-UHFFFAOYSA-N
12527
1-iodohexane
CCCCCCI

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4370g/mL
Boiling Point 179.0°C to 180.0°C
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4910 to 1.4930
Linear Formula CH3(CH2)5I
Quantity 100 mL
Beilstein 01, 146
Specific Gravity 1.437
Synonym hexyl iodide, n-hexyl iodide, hexane, 1-iodo, 1-hexyl iodide, iodohexane, 1-iodo-hexane, hexane,1-iodo, n-hexyliodide, iodo-hexane, 1-iodanylhexane
Solubility Information Solubility in water: insoluble
Stabilizer copper
Formula Weight 212.07
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Iodohexane, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective glov
EINECSNumber 211-339-

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit