Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Iodonaphthalene, 98%

Click to view available options
Quantity:
10g
50g
90-14-2
C10H7I
254.07
MFCD00003876
NHPPIJMARIVBGU-UHFFFAOYSA-N
naphthalene, 1-iodo,1-naphthyl iodide,alpha-iodonaphthalene,iodonaphthalene,1-iodonaphthalenen,.alpha.-iodonaphthalene,1-iodo-naphthalene,1-iodonaphthlen,1-iodonaphthlene,1-iodonapthalene
7004
1-iodonaphthalene
C1=CC=C2C(=C1)C=CC=C2I
90-14-2
C10H7I
254.07
MFCD00003876
NHPPIJMARIVBGU-UHFFFAOYSA-N
naphthalene, 1-iodo,1-naphthyl iodide,alpha-iodonaphthalene,iodonaphthalene,1-iodonaphthalenen,.alpha.-iodonaphthalene,1-iodo-naphthalene,1-iodonaphthlen,1-iodonaphthlene,1-iodonapthalene
7004
1-iodonaphthalene
C1=CC=C2C(=C1)C=CC=C2I
Applications
1-Iodonaphthalene undergoes Pd catalyzed cross-coupling reaction (Stille reaction) with 2,4-dimethoxy-6-(trimethylstannyl)pyrimidine to afford 2,4-dimethoxy-6-(naphthalen-1-yl)pyrimidine.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
Boiling Point 161°C to 162°C (15mmHg)
Quantity 50g
Melting Point 4°C
Flash Point >110°C (230°F)
Beilstein 1906415
Formula Weight 254.07
Percent Purity 98%
Chemical Name or Material 1-Iodonaphthalene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA