Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isocyanato-3-(phenoxymethyl)benzene, 97%, Thermo Scientific™

Catalog Number p-3522406
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
71219-80-2
C14H11NO2
225.25
MFCD09025914
CXYJFMVKSSTBDJ-UHFFFAOYSA-N
1-isocyanato-3-phenoxymethyl benzene, 3-phenoxymethyl benzenisocyanate, benzene,1-isocyanato-3-phenoxymethyl
18525913
1-isocyanato-3-(phenoxymethyl)benzene
O=C=NC1=CC=CC(COC2=CC=CC=C2)=C1
71219-80-2
C14H11NO2
225.25
MFCD09025914
CXYJFMVKSSTBDJ-UHFFFAOYSA-N
1-isocyanato-3-phenoxymethyl benzene, 3-phenoxymethyl benzenisocyanate, benzene,1-isocyanato-3-phenoxymethyl
18525913
1-isocyanato-3-(phenoxymethyl)benzene
O=C=NC1=CC=CC(COC2=CC=CC=C2)=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 225.25
Percent Purity 97%
Chemical Name or Material 1-Isocyanato-3-(phenoxymethyl)benzene
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit