Alfa Aesar™ 1-Isopropylpiperazine, 98%

Manufacturer: Alfa Aesar™ B2532506

Quantity 5g

 View more versions of this product

Catalog No. AAB2532506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4318-42-7
    Molecular Formula C7H16N2
    Formula Weight 128.22
    MDL Number MFCD00167971
    UN Number UN2733
    Chemical Name or Material 1-Isopropylpiperazine

    Specifications

    Formula Weight 128.22
    Molecular Formula C7H16N2
    MDL Number MFCD00167971
    CAS 4318-42-7
    UN Number UN2733
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H314-H318-H226
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
    Flash Point 54°C (130°F)
    Boiling Point 180° to 181°C
    Quantity 5g
    Chemical Name or Material 1-Isopropylpiperazine
    Assay 98%

    Description

    Applications
    1-Isopropylpiperazine is used as pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H311-H314-H318-H226
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit