Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isopropylpiperazine, 98+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
4318-42-7
C7H16N2
128.22
MFCD00167971
WHKWMTXTYKVFLK-UHFFFAOYSA-N
1-isopropylpiperazine,n-isopropylpiperazine,1-propan-2-yl piperazine,piperazine, 1-isopropyl,isopropyl piperazine,1-isopropyl-piperazine,1-1-methylethyl piperazine,n-isopropyl piperazine,1-isopropyl piperazine,piperazine, 1-1-methylethyl
78013
1-propan-2-ylpiperazine
CC(C)N1CCNCC1
4318-42-7
C7H16N2
128.22
MFCD00167971
WHKWMTXTYKVFLK-UHFFFAOYSA-N
1-isopropylpiperazine,n-isopropylpiperazine,1-propan-2-yl piperazine,piperazine, 1-isopropyl,isopropyl piperazine,1-isopropyl-piperazine,1-1-methylethyl piperazine,n-isopropyl piperazine,1-isopropyl piperazine,piperazine, 1-1-methylethyl
78013
1-propan-2-ylpiperazine
CC(C)N1CCNCC1
Boiling Point 180°C to 181°C
Flash Point 54°C
Refractive Index 1.4700 to 1.472
Specific Gravity 0.896
Color Yellow
Density 0.8960g/mL
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Formula Weight 128.22
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material 1-Isopropylpiperazine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 224-345-3