Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isoquinolinecarbonitrile, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
1198-30-7
C10H6N2
154.17
HJHXYSBRTVFEDD-UHFFFAOYSA-N
1-cyanoisoquinoline,1-isoquinolinecarbonitrile,isoquinaldonitrile,isoquinolinecarbonitrile,1-cyano-isoquinoline,isoquinaldinic nitrile,pubchem6269,maybridge1_002114,timtec-bb sbb002487
306057
isoquinoline-1-carbonitrile
C1=CC=C2C(=C1)C=CN=C2C#N
1198-30-7
C10H6N2
154.17
HJHXYSBRTVFEDD-UHFFFAOYSA-N
1-cyanoisoquinoline,1-isoquinolinecarbonitrile,isoquinaldonitrile,isoquinolinecarbonitrile,1-cyano-isoquinoline,isoquinaldinic nitrile,pubchem6269,maybridge1_002114,timtec-bb sbb002487
306057
isoquinoline-1-carbonitrile
C1=CC=C2C(=C1)C=CN=C2C#N
Quantity 5g
Melting Point 90.0°C to 92.0°C
Packaging Glass bottle
Assay Percent Range 97.5% min. (GC)
Formula Weight 154.17
Percent Purity 98%
Chemical Name or Material 1-Isoquinolinecarbonitrile

GHS Signal Word: Warning