Learn More
Andwin ScientificSupplier Diversity Partner 1 L WIDE MOUTH GLASS 12/CS DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner IR3410950
These I-CHEM Certified 300 Series Wide Mouth Jars come with readable bar-code lot label that can be used for convenient tracking. These jars are made to conform the US EPA Specifications and Guidance for Contaminant-Free Sample Containers.
Catalog No. 50-112-5987