Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methoxy-2-propanol, ≥99.7% (GC), MilliporeSigma™ Supelco™

Manufacturer:  MilliporeSigma Supelco 656921MLF

 View more versions of this product

Catalog No. 11-102-0586


Add to cart

Description

Description

Featured Industries: Environmental
Specifications

Specifications

107-98-2
MFCD00004537
90.12
≥99.7% (GC)
532°F
3.12 (vs. air)
118°C to 119°C
82°C - closed cup
CH3 CH(OH)CH2 OCH3
Propylene glycol methyl ether; Propyleneglycol monomethyl ether
Neat
Analytical Standard
10.9 mmHg (25°C)
0.916 g/mL (at 25°C)
1 mL
Analytical Standard
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit