Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methoxy-2-propanol Acetate, 97%, Spectrum™

Click to view available options
Quantity:
1 L
20 L
200 L
4 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
108-65-6
C6H12O3
132.16
LLHKCFNBLRBOGN-UHFFFAOYNA-N
1-methoxypropan-2-yl acetate
COCC(C)OC(C)=O
108-65-6
C6H12O3
132.16
LLHKCFNBLRBOGN-UHFFFAOYNA-N
1-methoxypropan-2-yl acetate
COCC(C)OC(C)=O

M2634, 108-65-6, C6H12O3

Spectrum™ 1-Methoxy-2-propanol Acetate
Packaging Drum
Quantity 200 L
CAS Max % 100%
UN Number UN3272
Percent Purity 97%