Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Methoxynaphthalene, 98+%

Click to view available options
Quantity:
25g
100g
500g
2216-69-5
C11H10O
158.2
MFCD00003924
NQMUGNMMFTYOHK-UHFFFAOYSA-N
naphthalene, 1-methoxy,methyl 1-naphthyl ether,1-methoxy naphthalene,naphthalene, methoxy,alpha-methoxynaphthalene,unii-dg2eol57lf,.alpha.-methoxynaphthalene,methoxynaphthalene,alpha-naphthyl methyl ether,dg2eol57lf
16668
1-methoxynaphthalene
COC1=CC=CC2=CC=CC=C21
2216-69-5
C11H10O
158.2
MFCD00003924
NQMUGNMMFTYOHK-UHFFFAOYSA-N
naphthalene, 1-methoxy,methyl 1-naphthyl ether,1-methoxy naphthalene,naphthalene, methoxy,alpha-methoxynaphthalene,unii-dg2eol57lf,.alpha.-methoxynaphthalene,methoxynaphthalene,alpha-naphthyl methyl ether,dg2eol57lf
16668
1-methoxynaphthalene
COC1=CC=CC2=CC=CC=C21
Applications
1-Methoxynaphthalene is used in the study of peroxygenase activity of cytochrome c peroxidase. It is used as a precursor to synthesize prenyl naphthalene-ols, which shows antioxidative activity.

Solubility
Immiscible with water. Miscible with ether, carbon disulfide and benzene.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Boiling Point 135°C to 137°C (12mmHg)
Melting Point 5°C
Flash Point >110°C (230°F)
Quantity 100g
Beilstein 7748884
Formula Weight 158.2
Percent Purity ≥98%
Chemical Name or Material 1-Methoxynaphthalene

RTECSNumber : QJ9465500

TSCA : TSCA