Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Methoxynaphthalene, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
1-Methoxynaphthalene is used in the study of peroxygenase activity of cytochrome c peroxidase. It is used as a precursor to synthesize prenyl naphthalene-ols, which shows antioxidative activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Methoxynaphthalene is used in the study of peroxygenase activity of cytochrome c peroxidase. It is used as a precursor to synthesize prenyl naphthalene-ols, which shows antioxidative activity.
Solubility
Immiscible with water. Miscible with ether, carbon disulfide and benzene.
Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
1-Methoxynaphthalene is used in the study of peroxygenase activity of cytochrome c peroxidase. It is used as a precursor to synthesize prenyl naphthalene-ols, which shows antioxidative activity.
Solubility
Immiscible with water. Miscible with ether, carbon disulfide and benzene.
Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 5°C |
Density | 1.094 |
Boiling Point | 135°C to 137°C (12 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.621 |
Quantity | 25 g |
Beilstein | 7748884 |
Solubility Information | Immiscible with water. Miscible with ether,carbon disulfide and benzene. |
Formula Weight | 158.2 |
Percent Purity | ≥98% |
Show More |
Safety and Handling
EINECSNumber | 218-696-1 |
RTECSNumber | QJ9465500 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction