Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1-cyclopentene 96%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
693-89-0
C6H10
82.13
MFCD00001397
ATQUFXWBVZUTKO-UHFFFAOYSA-N
1-methyl-1-cyclopentene,cyclopentene, 1-methyl,methylcyclopentene,cyclopentene, methyl,unii-v03m1dzc9i,1-methylcyclopent-1-ene,v03m1dzc9i,methyl-cyclopentene,1-methylcyclopentene-1,cyc1opentene,l-methyl
12746
1-methylcyclopentene
CC1=CCCC1
693-89-0
C6H10
82.13
MFCD00001397
ATQUFXWBVZUTKO-UHFFFAOYSA-N
1-methyl-1-cyclopentene,cyclopentene, 1-methyl,methylcyclopentene,cyclopentene, methyl,unii-v03m1dzc9i,1-methylcyclopent-1-ene,v03m1dzc9i,methyl-cyclopentene,1-methylcyclopentene-1,cyc1opentene,l-methyl
12746
1-methylcyclopentene
CC1=CCCC1
Density 0.7800g/mL
Refractive Index 1.4310 to 1.433
Flash Point -17°C
Boiling Point 72°C (754.0mmHg)
Specific Gravity 0.78
Infrared Spectrum Authentic
Quantity 25mL
Packaging Glass bottle
Color Colorless
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula C5H7CH3
Beilstein 05, 64
Formula Weight 82.13
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material 1-Methyl-1-cyclopentene, 96%
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-762-0

TSCA : TSCA