Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1-phenylhydrazine, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
618-40-6
C7H10N2
122.17
MFCD00007627
MWOODERJGVWYJE-UHFFFAOYSA-N
1-methylphenylhydrazine,hydrazine, 1-methyl-1-phenyl,n-methyl-n-phenylhydrazine,n-phenyl-n-methylhydrazine,unii-yhb9czb63u,1-methyl-1-phenyl-hydrazine,alpha-methyl-alpha-phenylhydrazine,hydrazine, methylphenyl,yhb9czb63u,.alpha.-methyl-.alpha.-phenylhydrazine
36299
1-methyl-1-phenylhydrazine
CN(C1=CC=CC=C1)N
618-40-6
C7H10N2
122.17
MFCD00007627
MWOODERJGVWYJE-UHFFFAOYSA-N
1-methylphenylhydrazine,hydrazine, 1-methyl-1-phenyl,n-methyl-n-phenylhydrazine,n-phenyl-n-methylhydrazine,unii-yhb9czb63u,1-methyl-1-phenyl-hydrazine,alpha-methyl-alpha-phenylhydrazine,hydrazine, methylphenyl,yhb9czb63u,.alpha.-methyl-.alpha.-phenylhydrazine
36299
1-methyl-1-phenylhydrazine
CN(C1=CC=CC=C1)N
Flash Point 96°C
Boiling Point 75°C to 78°C (0.4 kPa)
Specific Gravity 1.04
Infrared Spectrum Authentic
Density 1.0400g/mL
Refractive Index 1.5810 to 1.585
Packaging Glass bottle
Color Undesignated
Quantity 5g
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Linear Formula C6H5N(CH3)NH2
Beilstein 15, 117
Solubility Information Solubility in water: sparingly soluble in water.,miscible in ethanol,ether,benzene,chloroform
Formula Weight 122.17
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 1-Methyl-1-phenylhydrazine, 96%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-547-9

TSCA : TSCA