Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Methyl-1-phenylhydrazine, 97%

Click to view available options
Quantity:
5g
25g
100g
618-40-6
C7H10N2
122.171
MFCD00007627
MWOODERJGVWYJE-UHFFFAOYSA-N
1-methylphenylhydrazine,hydrazine, 1-methyl-1-phenyl,n-methyl-n-phenylhydrazine,n-phenyl-n-methylhydrazine,unii-yhb9czb63u,1-methyl-1-phenyl-hydrazine,alpha-methyl-alpha-phenylhydrazine,hydrazine, methylphenyl,yhb9czb63u,.alpha.-methyl-.alpha.-phenylhydrazine
36299
1-methyl-1-phenylhydrazine
CN(C1=CC=CC=C1)N
618-40-6
C7H10N2
122.171
MFCD00007627
MWOODERJGVWYJE-UHFFFAOYSA-N
1-methylphenylhydrazine,hydrazine, 1-methyl-1-phenyl,n-methyl-n-phenylhydrazine,n-phenyl-n-methylhydrazine,unii-yhb9czb63u,1-methyl-1-phenyl-hydrazine,alpha-methyl-alpha-phenylhydrazine,hydrazine, methylphenyl,yhb9czb63u,.alpha.-methyl-.alpha.-phenylhydrazine
36299
1-methyl-1-phenylhydrazine
CN(C1=CC=CC=C1)N
Applications
1-Methyl-1-phenylhydrazine is used to make other chemicals.

Solubility
Slightly soluble in water.

Notes
Storage temperature is 2 - 8°C. Incompatible with oxidizing agents.
Flash Point 92°C (198°F)
Quantity 100g
Boiling Point 54°C to 55°C (0.3mmHg)
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 1-Methyl-1-phenylhydrazine

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MV7712000

TSCA : TSCA