Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Acros Organics

missing translation for 'orderingAttributeHoverText'
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
85676-85-3,109-99-9
C4H7BrMg
159.31
FGYXQBPXHQHYNU-UHFFFAOYSA-M
1-methyl-1-propenylmagnesium bromide,1-methyl-1-propenyl magnesium bromide,1-methylpropenylmagnesium bromide,fejkwduhnfcdrz-uhfffaoysa-m,e-but-2-en-2-ylmagnesium bromide,magnesium, bromo 1-methyl-1-propenyl,bromo 2e-but-2-en-2-yl magnesium,bromo 2z-but-2-en-2-yl magnesium,bromo 1e-1-methyl-1-propen-1-yl magnesium,1-methyl-1-propenylmagnesium bromide solution, 0.5 m in thf
3597816
magnesium;but-2-ene;bromide
CC=[C-]C.[Mg+2].[Br-]
85676-85-3,109-99-9
C4H7BrMg
159.31
FGYXQBPXHQHYNU-UHFFFAOYSA-M
1-methyl-1-propenylmagnesium bromide,1-methyl-1-propenyl magnesium bromide,1-methylpropenylmagnesium bromide,fejkwduhnfcdrz-uhfffaoysa-m,e-but-2-en-2-ylmagnesium bromide,magnesium, bromo 1-methyl-1-propenyl,bromo 2e-but-2-en-2-yl magnesium,bromo 2z-but-2-en-2-yl magnesium,bromo 1e-1-methyl-1-propen-1-yl magnesium,1-methyl-1-propenylmagnesium bromide solution, 0.5 m in thf
3597816
magnesium;but-2-ene;bromide
CC=[C-]C.[Mg+2].[Br-]
Density 0.9340g/mL
Boiling Point 67.0°C
Specific Gravity 0.934
Color Colorless to Yellow
Quantity 800mL
Packaging AcroSeal™ Glass Bottle
Flash Point −26°C
CAS Min % 7.68,90.62
CAS Max % 9.38,92.32
Solubility Information Solubility in water: reacts
Formula Weight 159.31
Physical Form Liquid
Chemical Name or Material 1-Methyl-1-propenylmagnesium bromide
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
In contact with water releases flammable gases.
Reacts violently with water.
May form explo

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a dry place.
Store in a closed conta

GHS Signal Word: Danger