Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1H-indazole-4-boronic acid pinacol ester, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
885698-94-2
C14H19BN2O2
258.13
OZHFBNULPUBRJF-UHFFFAOYSA-N
1-methyl-1h-indazole-4-boronic acid pinacol ester,1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole,1-methylindazole-4-boronic acid pinacol ester,1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole,1-methyl-1h-indazole-4-boronic acid, pinacol ester,1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl indazole,1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole,1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indazole,1h-indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
45480187
1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole
B1(OC(C(O1)(C)C)(C)C)C2=C3C=NN(C3=CC=C2)C
885698-94-2
C14H19BN2O2
258.13
OZHFBNULPUBRJF-UHFFFAOYSA-N
1-methyl-1h-indazole-4-boronic acid pinacol ester,1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole,1-methylindazole-4-boronic acid pinacol ester,1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole,1-methyl-1h-indazole-4-boronic acid, pinacol ester,1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl indazole,1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole,1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indazole,1h-indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
45480187
1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole
B1(OC(C(O1)(C)C)(C)C)C2=C3C=NN(C3=CC=C2)C
Melting Point 83.0°C to 86.0°C
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 258.13
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indazole-4-boronic acid pinacol ester

GHS Signal Word: Warning