Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1H-pyrazole-4-boronic acid hydrochloride, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
1026796-02-0
C4H7BN2O2·ClH
162.38
YFPLZINTZVVVJM-UHFFFAOYSA-N
1-methylpyrazole-4-boronic acid, hcl,1-methyl-1h-pyrazol-4-yl boronic acid hydrochloride,1-methyl-1h-pyrazole-4-boronic acid hydrochloride,1-methylpyrazole-4-boronic acid hcl,1-methyl-1h-pyrazol-4-ylboronic acid hydrochloride,1-methylpyrazol-4-ylboronic acid hydrochloride,1-methylpyrazole-4-boronic acid,hcl,1-methyl-1h-pyrazole-4-boronic acid xhcl,1-methylpyrazole-4-boronic acid hydrochloride,1-methyl-1h-pyrazole-4-boronic acid hcl
45158882
(1-methylpyrazol-4-yl)boronic acid;hydrochloride
B(C1=CN(N=C1)C)(O)O.Cl
1026796-02-0
C4H7BN2O2·ClH
162.38
YFPLZINTZVVVJM-UHFFFAOYSA-N
1-methylpyrazole-4-boronic acid, hcl,1-methyl-1h-pyrazol-4-yl boronic acid hydrochloride,1-methyl-1h-pyrazole-4-boronic acid hydrochloride,1-methylpyrazole-4-boronic acid hcl,1-methyl-1h-pyrazol-4-ylboronic acid hydrochloride,1-methylpyrazol-4-ylboronic acid hydrochloride,1-methylpyrazole-4-boronic acid,hcl,1-methyl-1h-pyrazole-4-boronic acid xhcl,1-methylpyrazole-4-boronic acid hydrochloride,1-methyl-1h-pyrazole-4-boronic acid hcl
45158882
(1-methylpyrazol-4-yl)boronic acid;hydrochloride
B(C1=CN(N=C1)C)(O)O.Cl
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 94% min. (HPLC)
Formula Weight 162.38
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-pyrazole-4-boronic acid hydrochloride

GHS Signal Word: Warning