Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester, 95%

Manufacturer:  Alfa Aesar™ H2770303

Quantity 1g

 View more versions of this product

Catalog No. AAH2770303


  Check Availability
Add to cart

Specifications

Description

Applications
Important intermediates and reagents in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

1044851-76-4
194.04
5-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-methyl-1H-pyrazole
C9H15BN2O2
MFCD07777079
1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester

Specifications

1044851-76-4
194.04
5-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-methyl-1H-pyrazole
1g
95%
C9H15BN2O2
MFCD07777079
72° to 75°C
1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit