Alfa Aesar™ 1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester, 95%

Manufacturer: Alfa Aesar™ H27703MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27703MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1044851-76-4
    Molecular Formula C9H15BN2O2
    Formula Weight 194.04
    MDL Number MFCD07777079
    Synonym 5-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-methyl-1H-pyrazole
    Chemical Name or Material 1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester

    Specifications

    Formula Weight 194.04
    Molecular Formula C9H15BN2O2
    MDL Number MFCD07777079
    CAS 1044851-76-4
    Synonym 5-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-methyl-1H-pyrazole
    Melting Point 72° to 75°C
    Quantity 250mg
    Chemical Name or Material 1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester
    Assay 95%

    Description

    Applications
    Important intermediates and reagents in organic synthesis.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit