Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Methyl-1H-pyrazole-5-boronic acid pinacol ester, 98%

Manufacturer:  Alfa Aesar™ H2709403

Quantity 1g

 View more versions of this product

Catalog No. AAH2709403


  Check Availability
Add to cart

Specifications

Description

Applications
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester is used as a pharmaceutical intermediate, agrochemical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Description & Specifications

Chemical Identifiers

847818-74-0
208.07
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
C10H17BN2O2
MFCD05861380
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester

Specifications

847818-74-0
208.07
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
1g
98%
C10H17BN2O2
MFCD05861380
58° to 61°C
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit