Alfa Aesar™ 1-Methyl-1H-pyrazole-5-boronic acid pinacol ester, 98%

Manufacturer: Alfa Aesar™ H2709406

Quantity 5g

 View more versions of this product

Catalog No. AAH2709406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 847818-74-0
    Molecular Formula C10H17BN2O2
    Formula Weight 208.07
    MDL Number MFCD05861380
    Synonym 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
    Chemical Name or Material 1-Methyl-1H-pyrazole-5-boronic acid pinacol ester

    Specifications

    Formula Weight 208.07
    Molecular Formula C10H17BN2O2
    MDL Number MFCD05861380
    Synonym 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
    Melting Point 58° to 61°C
    Quantity 5g
    CAS 847818-74-0
    Chemical Name or Material 1-Methyl-1H-pyrazole-5-boronic acid pinacol ester
    Assay 98%

    Description

    Applications
    1-Methyl-1H-pyrazole-5-boronic acid pinacol ester is used as a pharmaceutical intermediate, agrochemical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit