Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl 2-Aminoterephthalate 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
60728-41-8
C9H9NO4
195.174
MFCD00189374
QKOKLMFCKLEFDV-UHFFFAOYSA-N
3-Amino-4-carbomethoxybenzoic Acid, 3-Amino-4-methoxycarbonylbenzoic Acid, 2-Aminoterephthalic Acid 1-Methyl Ester
5235500
3-amino-4-methoxycarbonylbenzoic acid
COC(=O)C1=C(C=C(C=C1)C(=O)O)N
60728-41-8
C9H9NO4
195.174
MFCD00189374
QKOKLMFCKLEFDV-UHFFFAOYSA-N
3-Amino-4-carbomethoxybenzoic Acid, 3-Amino-4-methoxycarbonylbenzoic Acid, 2-Aminoterephthalic Acid 1-Methyl Ester
5235500
3-amino-4-methoxycarbonylbenzoic acid
COC(=O)C1=C(C=C(C=C1)C(=O)O)N
Melting Point 218°C
Color Yellow
Quantity 25g
Formula Weight 195.17
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 1-Methyl 2-Aminoterephthalate