Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-2-imidazolecarboxaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
13750-81-7
C5H6N2O
110.12
MFCD01321308
UEBFLTZXUXZPJO-UHFFFAOYSA-N
1-methyl-1h-imidazole-2-carbaldehyde,1-methyl-2-imidazolecarboxaldehyde,1-methylimidazole-2-carboxaldehyde,1h-imidazole-2-carboxaldehyde, 1-methyl,2-formyl-1-methylimidazole,imidazole-2-carboxaldehyde, 1-methyl,pubchem7614,zlchem 1231,2-formyl-l-methylimidazole,acmc-209ce6
139575
1-methylimidazole-2-carbaldehyde
CN1C=CN=C1C=O
13750-81-7
C5H6N2O
110.12
MFCD01321308
UEBFLTZXUXZPJO-UHFFFAOYSA-N
1-methyl-1h-imidazole-2-carbaldehyde,1-methyl-2-imidazolecarboxaldehyde,1-methylimidazole-2-carboxaldehyde,1h-imidazole-2-carboxaldehyde, 1-methyl,2-formyl-1-methylimidazole,imidazole-2-carboxaldehyde, 1-methyl,pubchem7614,zlchem 1231,2-formyl-l-methylimidazole,acmc-209ce6
139575
1-methylimidazole-2-carbaldehyde
CN1C=CN=C1C=O
Boiling Point 70°C to 74°C (1.0mmHg)
Quantity 5g
Flash Point 101°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
Melting Point 35°C to 39°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 110.12
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1-Methyl-2-imidazolecarboxaldehyde
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning