Alfa Aesar™ 1-Methyl-2-phenylindole, 99%

Manufacturer: Alfa Aesar™ B2210530

Quantity 250g

 View more versions of this product

Catalog No. AAB2210530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3558-24-5
    Molecular Formula C15H13N
    Formula Weight 207.28
    MDL Number MFCD00022892
    Beilstein 148770
    Chemical Name or Material 1-Methyl-2-phenylindole

    Specifications

    Formula Weight 207.28
    TSCA TSCA
    Molecular Formula C15H13N
    MDL Number MFCD00022892
    CAS 3558-24-5
    Beilstein 148770
    Melting Point 98° to 99°C
    Quantity 250g
    Chemical Name or Material 1-Methyl-2-phenylindole
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit