Alfa Aesar™ 1-Methyl-2-pyridone, 99+%

Manufacturer: Alfa Aesar™ B2424414

Quantity 25g

 View more versions of this product

Catalog No. AAB2424414

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 694-85-9
    Molecular Formula C6H7NO
    Formula Weight 109.13
    MDL Number MFCD00006520
    Beilstein 107805
    Chemical Name or Material 1-Methyl-2-pyridone

    Specifications

    Formula Weight 109.13
    Boiling Point 249° to 251°C
    TSCA TSCA
    Molecular Formula C6H7NO
    MDL Number MFCD00006520
    CAS 694-85-9
    Beilstein 107805
    Flash Point >110°C (230°F)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number UV1148100
    Quantity 25g
    Chemical Name or Material 1-Methyl-2-pyridone
    Assay ≥99%

    Description

    Applications
    1-Methyl-2-pyridone is used in pharmaceutical industry. A common problem with hydrophobic non-natural base pairs—the inefficient extension following such a base pair—is reduced by using the methylpyridone scaffold, which simultaneously limits another frequent problem of non-natural bases: mispairs with deoxyadenosine.

    Solubility
    Soluble in water (1000 mg/ml at 20°C).

    Notes
    Store at room temperature. Incompatible with oxidizing agents.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit