Learn More
1-Methyl-2-pyrrolidinone, 99+%, Thermo Scientific Chemicals
Description
1-methyl-2-pyrrolidinone is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility. It is used as stripping agent, oven cleaners and paint strippers. It is involved in the extraction of lube oil process and in the purification of synthetic gas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-methyl-2-pyrrolidinone is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility. It is used as stripping agent, oven cleaners and paint strippers. It is involved in the extraction of lube oil process and in the purification of synthetic gas.
Solubility
Miscible with water, lower alcohols, ketones, ethyl acetate, castor oil, chloroform and benzene. Moderately miscible with aliphatic hydrocarbons.
Notes
Light sensitive and hygroscopic. Incompatible with strong oxidizing agents. Hygroscopic.
Specifications
Melting Point | -24°C |
Density | 1.033 |
Boiling Point | 204°C |
Quantity | 500 g |
Flash Point | 86°C (187°F) |
Odor | Amine-like |
Refractive Index | 1.4705 |
Beilstein | 106420 |
Solubility Information | Miscible with water,lower alcohols,ketones,ethyl acetate,castor oil,chloroform and benzene. Moderately miscible with aliphatic hydrocarbons. |
Formula Weight | 99.13 |
Show More |
Safety and Handling
Hazard Category | H227-H315-H319-H335-H360D-H373 |
Precautionary Statement | P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c |
EINECSNumber | 212-828-1 |
RTECSNumber | UY5790000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.