Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals

Catalog Number AC364381000
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
872-50-4
C5H9NO
99.133
MFCD00003193
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
13387
CHEBI:7307
1-methylpyrrolidin-2-one
CN1CCCC1=O
872-50-4
C5H9NO
99.133
MFCD00003193
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
13387
CHEBI:7307
1-methylpyrrolidin-2-one
CN1CCCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -24.0°C
Color Colorless
CAS Min % 99.4
CAS Max % 100.0
Density 1.0300g/mL
Boiling Point 202.0°C
Quantity 100 mL
Flash Point 91°C
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Refractive Index 1.4690 to 1.4710
Assay Percent Range 99.5%
Specific Gravity 1.03
Viscosity 1.67 mPa.s (20°C)
Beilstein 21, 237
Water 0.005% Max. (Coulometric)
Fieser 01,696; 02,281; 09,316; 11,346
Merck Index 15, 6190
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 99.13
Percent Purity 99.5%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material 1-Methyl-2-pyrrolidinone, Over Molecular Sieves
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May damage the unborn child.
Precautionary Statement GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-828-1
RTECSNumber UY5790000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit