Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-2-pyrrolidinone, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
25L
Packaging:
Glass Bottle
Metal drum
872-50-4
C5H9NO
99.133
MFCD00003193
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methyl-2-pyrrolidinone,n-methyl-2-pyrrolidone,n-methylpyrrolidone,1-methyl-2-pyrrolidone,methylpyrrolidone,m-pyrol,n-methyl-2-pyrrolidinone,n-methylpyrrolidinone,1-methylpyrrolidinone,2-pyrrolidinone, 1-methyl
13387
CHEBI:7307
1-methylpyrrolidin-2-one
CN1CCCC1=O
872-50-4
C5H9NO
99.133
MFCD00003193
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methyl-2-pyrrolidinone,n-methyl-2-pyrrolidone,n-methylpyrrolidone,1-methyl-2-pyrrolidone,methylpyrrolidone,m-pyrol,n-methyl-2-pyrrolidinone,n-methylpyrrolidinone,1-methylpyrrolidinone,2-pyrrolidinone, 1-methyl
13387
CHEBI:7307
1-methylpyrrolidin-2-one
CN1CCCC1=O
CAS Min % 98.5
CAS Max % 100.0
Additional Information Vapor Pressure: 0.7mbar at 25°C
Assay Percent Range 99%
Beilstein 21, 237
Fieser 01,696; 02,281; 09,316; 11,346
Merck Index 15, 6190
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 99.13
Physical Form Liquid
Percent Purity 99%
Grade Extra Pure
Density 1.0300g/mL
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.4690 to 1.4710
Specific Gravity 1.03
Water 0.05% max. (K.F.)
Flash Point 91°C
Viscosity 1.67 mPa.s (20°C)
Boiling Point 202.0°C
Color Undesignated
Melting Point -24.0°C
Quantity 2.5L
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May damage the unborn child.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger