Alfa Aesar™ 1-Methyl-2-pyrrolidinone, ACS grade, 99.0+%

Manufacturer: Alfa Aesar™ 43894K7

Quantity 4L

 View more versions of this product

Catalog No. AA43894K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 872-50-4
    Molecular Formula C5H9NO
    Formula Weight 99.13
    MDL Number MFCD00003193
    Beilstein 106420
    Merck Index 14,6117
    Chemical Name or Material 1-Methyl-2-pyrrolidinone
    Grade ACS Grade

    Specifications

    Formula Weight 99.13
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C5H9NO
    Density 1.033g/mL
    Physical Form Liquid
    CAS 872-50-4
    Beilstein 106420
    Flash Point 86°C (187°F)
    Health Hazard 2 GHS H Statement
    H360-H315-H319-H335-H227
    May damage fertility or the unborn child.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    TSCA TSCA
    MDL Number MFCD00003193
    Boiling Point 204°C
    Health Hazard 1 Danger
    Quantity 4L
    Odor Amine-like
    Melting Point −24°C
    Color Colorless to pale yellow
    Sensitivity Light Sensitive & Hygroscopic
    Refractive Index 1.4705
    Merck Index 14,6117
    RTECS Number UY5790000
    Chemical Name or Material 1-Methyl-2-pyrrolidinone
    Assay ≥99.0%
    Grade ACS Grade

    GHS H Statement
    H360-H315-H319-H335-H227
    May damage fertility or the unborn child.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit