Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Methyl-2-pyrrolidinone, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 43741AE

Quantity 100mL

 View more versions of this product

Catalog No. AA43741AE


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

872-50-4
99.13
106420
1-Methyl-2-pyrrolidinone, Anhydrous
C5H9NO
MFCD00003193
14,6117

Specifications

Light Sensitive & Hygroscopic
872-50-4
Amine-like
1.4705
C5H9NO
MFCD00003193
14,6117
Colorless to pale yellow
86°C (187°F)
99.5%
100mL
1.033g/mL
ChemSeal™ bottles
Liquid
99.13
106420
−24°C
204°C
1-Methyl-2-pyrrolidinone, Anhydrous

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360-H315-H319-H335-H227
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : UY5790000

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit