Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Methyl-2-pyrrolidinone-d9, 97% (Isotopic)

NMR Solvent

Manufacturer:  Alfa Aesar™ 4233603

Quantity 1g

 View more versions of this product

Catalog No. AA4233603


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

185964-60-7
108.21
MFCD00148961
1-Methyl-2-pyrrolidinone-d9, Isotopic

Specifications

185964-60-7
−24°C
1g
86°C (187°F)
MFCD00148961
97%
81° to 82°C (10mmHg)
Liquid
Colorless
108.21
1-Methyl-2-pyrrolidinone-d9, Isotopic

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360-H315-H319-H335-H227
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P201-P309-P310
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit