Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-2-pyrrolidinone, HPLC Grade, 99.5%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
Packaging:
Amber Glass Bottle
872-50-4
C5H9NO
99.13
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methylpyrrolidin-2-one
CN1CCCC1=O
872-50-4
C5H9NO
99.13
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methylpyrrolidin-2-one
CN1CCCC1=O

HP772, 872-50-4, C5H9NO

Spectrum™ 1-Methyl-2-pyrrolidinone, HPLC Grade is a laboratory analytical reagent that is miscible with water and several other solvents, including ethyl acetate, chloroform, benzene and alcohols and ketones. All HPLC and Ultrapure solvents offered by Spectrum are glass distilled, submicron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.
CAS Max % 100%
Percent Purity 99.5%
Packaging Amber Glass Bottle
Residue after Ignition 10 ppm
Titratable Base 1 μeq/g
Water 0.08%
Quantity 4 L