Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Methyl-2-pyrrolidinone, HPLC Grade, 99.5%, Spectrum™ Chemical

Click to view available options
Quantity:
1 L
4 x 4 L
6 x 1 L
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical 1-Methyl-2-pyrrolidinone, HPLC Grade is a laboratory analytical reagent that is miscible with water and several other solvents, including ethyl acetate, chloroform, benzene and alcohols and ketones. All HPLC and Ultrapure solvents offered by Spectrum are glass distilled, submicron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.
Specifications
CAS Max % | 100% |
Percent Purity | 99.5% |
Quantity | 4 x 4 L |
Packaging | Amber Glass Bottle |
Residue after Ignition | 10 ppm |
Titratable Base | 1 μeq/g |
Water | 0.08% |