Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-2-pyrrolidone, ACS, 99%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
872-50-4
872-50-4
CAS Max % 1
Percent Purity 0.99
Grade ACS
Chloride 1 ppm
Packaging Amber Glass Bottle
Water 0.0005
Quantity 4 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.