Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3,4-dihydroisoquinoline hydrochloride hydrate, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
26210-39-9
C10H12ClN
181.663
MFCD08059359
PGUZPOADJHTSNH-UHFFFAOYSA-N
1-methyl-3,4-dihydroisoquinoline hydrochloride,1-methyl-3,4-dihydroisoquinoline, chloride
2724663
1-methyl-3,4-dihydroisoquinoline;hydrochloride
CC1=NCCC2=CC=CC=C12.Cl
26210-39-9
C10H12ClN
181.663
MFCD08059359
PGUZPOADJHTSNH-UHFFFAOYSA-N
1-methyl-3,4-dihydroisoquinoline hydrochloride,1-methyl-3,4-dihydroisoquinoline, chloride
2724663
1-methyl-3,4-dihydroisoquinoline;hydrochloride
CC1=NCCC2=CC=CC=C12.Cl
Quantity 5g
Packaging Glass bottle
Formula Weight 181.66
Chemical Name or Material 1-Methyl-3, 4-dihydroisoquinoline hydrochloride hydrate

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.