Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3,6-(1H,2H)-pyridazinedione, Acros Organics

Click to view available options
Quantity:
25g
5436-01-1
C5H6N2O2
126.12
MFCD00234108
UAECOHJYXUJDOF-UHFFFAOYSA-N
3-hydroxy-1-methylpyridazin-6 1h-one,1-methyl-3,6-pyridazinedione,n-methyl-3,6-pyridazinedione,n-methylmaleic hydrazide,6-hydroxy-2-methylpyridazin-3 2h-one,n-methyl maleic hydrazide,1-methyl-3,6-1h,2h pyridazinedione,1-methyl-1,2-dihydropyridazine-3,6-dione,6-hydroxy-2-methylpyridazin-3-one,1-methyl-1,2-dihydro-3,6-pyridazinedione
95157
2-methyl-1H-pyridazine-3,6-dione
CN1C(=O)C=CC(=O)N1
5436-01-1
C5H6N2O2
126.12
MFCD00234108
UAECOHJYXUJDOF-UHFFFAOYSA-N
3-hydroxy-1-methylpyridazin-6 1h-one,1-methyl-3,6-pyridazinedione,n-methyl-3,6-pyridazinedione,n-methylmaleic hydrazide,6-hydroxy-2-methylpyridazin-3 2h-one,n-methyl maleic hydrazide,1-methyl-3,6-1h,2h pyridazinedione,1-methyl-1,2-dihydropyridazine-3,6-dione,6-hydroxy-2-methylpyridazin-3-one,1-methyl-1,2-dihydro-3,6-pyridazinedione
95157
2-methyl-1H-pyridazine-3,6-dione
CN1C(=O)C=CC(=O)N1
Quantity 25g
Formula Weight 126.12
Chemical Name or Material 1-Methyl-3, 6-(1H, 2H)-pyridazinedione