Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-indoleacetic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1912-48-7
C11H11NO2
189.21
NAIPEFIYIQFVFC-UHFFFAOYSA-N
1-methyl-3-indoleacetic acid,2-1-methyl-1h-indol-3-yl acetic acid,1h-indole-3-acetic acid, 1-methyl,2-1-methylindol-3-yl acetic acid,1-methylindole-3-acetic acid,1-methyl-3-indolyl acetic acid,1-methylindol-3-yl acetic acid,1-methyl-1h-indole-3-acetic acid,1-methyl-3-indoleaceticacid,1-methyl-1 h-indol-3-yl acetic acid
254166
2-(1-methylindol-3-yl)acetic acid
CN1C=C(C2=CC=CC=C21)CC(=O)O
1912-48-7
C11H11NO2
189.21
NAIPEFIYIQFVFC-UHFFFAOYSA-N
1-methyl-3-indoleacetic acid,2-1-methyl-1h-indol-3-yl acetic acid,1h-indole-3-acetic acid, 1-methyl,2-1-methylindol-3-yl acetic acid,1-methylindole-3-acetic acid,1-methyl-3-indolyl acetic acid,1-methylindol-3-yl acetic acid,1-methyl-1h-indole-3-acetic acid,1-methyl-3-indoleaceticacid,1-methyl-1 h-indol-3-yl acetic acid
254166
2-(1-methylindol-3-yl)acetic acid
CN1C=C(C2=CC=CC=C21)CC(=O)O
Quantity 1g
Packaging Glass bottle
Melting Point 125.0°C to 126.5°C
Color Tan to White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 189.21
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-Methyl-3-indoleacetic acid
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED: rinse mouth.
Do

GHS Signal Word: Danger