Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-octylimidazolium chloride, 97%, Acros Organics

Click to view available options
Quantity:
5g
64697-40-1
C12H23ClN2
230.78
OXFBEEDAZHXDHB-UHFFFAOYSA-M
1-methyl-3-n-octylimidazolium chloride,1-methyl-3-octylimidazolium chloride,1-octyl-3-methylimidazolium chloride,3-methyl-1-octylimidazolium chloride,omimcl,acmc-20aj1j,dsstox_cid_27849,dsstox_rid_82602,dsstox_gsid_47873,ksc491c7j
2734223
CHEBI:61342
1-methyl-3-octylimidazol-1-ium;chloride
CCCCCCCCN1C=C[N+](=C1)C.[Cl-]
64697-40-1
C12H23ClN2
230.78
OXFBEEDAZHXDHB-UHFFFAOYSA-M
1-methyl-3-n-octylimidazolium chloride,1-methyl-3-octylimidazolium chloride,1-octyl-3-methylimidazolium chloride,3-methyl-1-octylimidazolium chloride,omimcl,acmc-20aj1j,dsstox_cid_27849,dsstox_rid_82602,dsstox_gsid_47873,ksc491c7j
2734223
CHEBI:61342
1-methyl-3-octylimidazol-1-ium;chloride
CCCCCCCCN1C=C[N+](=C1)C.[Cl-]
Ionic liquid
Specific Gravity 1.0124
Quantity 5g
Density 1.0124g/mL
Refractive Index 1.5060 to 1.5080
Assay Percent Range 97%
Formula Weight 230.78
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-octylimidazolium chloride