Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-phenylpiperazine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
5271-27-2
C11H16N2
176.263
MFCD03411603
IRMBVBDXXYXPEW-UHFFFAOYSA-N
1-methyl-3-phenyl-piperazine,1-n-methyl-3-phenyl-piperazine,1-methyl-3-phenyl piperazine,piperazine, 1-methyl-3-phenyl,+/--3-phenyl-1-methylpiperazine,dl-1-methyl-3-phenyl-piperazine,1-methyl-3-phenyl-1-piperazine,pubchem8578,3-phenyl-1-methylpiperazine, +/-,n-methyl-3-phenylpiperazine
2760009
1-methyl-3-phenylpiperazine
CN1CCNC(C1)C2=CC=CC=C2
5271-27-2
C11H16N2
176.263
MFCD03411603
IRMBVBDXXYXPEW-UHFFFAOYSA-N
1-methyl-3-phenyl-piperazine,1-n-methyl-3-phenyl-piperazine,1-methyl-3-phenyl piperazine,piperazine, 1-methyl-3-phenyl,+/--3-phenyl-1-methylpiperazine,dl-1-methyl-3-phenyl-piperazine,1-methyl-3-phenyl-1-piperazine,pubchem8578,3-phenyl-1-methylpiperazine, +/-,n-methyl-3-phenylpiperazine
2760009
1-methyl-3-phenylpiperazine
CN1CCNC(C1)C2=CC=CC=C2
Melting Point 55.0°C to 59.0°C
Quantity 5g
Infrared Spectrum Authentic
Color Yellow
Flash Point >110°C
Packaging Glass bottle
Boiling Point 85.0°C (0.5 mmHg)
Assay Percent Range 99%
Formula Weight 176.26
Physical Form Needles or Crystals
Percent Purity 99%
Chemical Name or Material 1-Methyl-3-phenylpiperazine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger