Alfa Aesar™ 1-Methyl-3-trifluoromethyl-2-pyrazolin-5-one, 96%

Manufacturer: Alfa Aesar™ A1189014

Quantity 25g

 View more versions of this product

Catalog No. AAA1189014

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 1481-02-3
    Molecular Formula C5H5F3N2O
    Formula Weight 166.1
    MDL Number MFCD00051654
    Beilstein 5811070
    Synonym 2,4-Dihydro-2-methyl-5-trifluoromethyl-3H-pyrazol-3-one
    Chemical Name or Material 1-Methyl-3-trifluoromethyl-2-pyrazolin-5-one

    Specifications

    Formula Weight 166.1
    Synonym 2,4-Dihydro-2-methyl-5-trifluoromethyl-3H-pyrazol-3-one
    Molecular Formula C5H5F3N2O
    CAS 1481-02-3
    Beilstein 5811070
    Melting Point 178° to 180°C
    Quantity 25g
    MDL Number MFCD00051654
    Chemical Name or Material 1-Methyl-3-trifluoromethyl-2-pyrazolin-5-one
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit