Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-4-iodo-1H-pyrazole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
39806-90-1
C4H5IN2
208
MFCD05663860
RSDRDHPLXWMTRJ-UHFFFAOYSA-N
4-iodo-1-methyl-1h-pyrazole,1-methyl-4-iodo-1h-pyrazole,1h-pyrazole, 4-iodo-1-methyl,1-methyl-4-iodopyrazole,zlchem 32,pubchem24346,acmc-1ctkh,n-methyl-4-iodopyrazole,1-methyl-4-iodo pyrazole,4-iodo-1-methyl-pyrazole
2734773
4-iodo-1-methylpyrazole
CN1C=C(C=N1)I
39806-90-1
C4H5IN2
208
MFCD05663860
RSDRDHPLXWMTRJ-UHFFFAOYSA-N
4-iodo-1-methyl-1h-pyrazole,1-methyl-4-iodo-1h-pyrazole,1h-pyrazole, 4-iodo-1-methyl,1-methyl-4-iodopyrazole,zlchem 32,pubchem24346,acmc-1ctkh,n-methyl-4-iodopyrazole,1-methyl-4-iodo pyrazole,4-iodo-1-methyl-pyrazole
2734773
4-iodo-1-methylpyrazole
CN1C=C(C=N1)I
Quantity 5g
Color White
Melting Point 61.0°C to 67.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 208
Physical Form Crystals or Crystalline Needles
Percent Purity 97%
Chemical Name or Material 1-Methyl-4-iodo-1H-pyrazole
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning