Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-4-piperidinemethanol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
20691-89-8
C7H15NO
129.2
KJZLJGZZDNGGCA-UHFFFAOYSA-N
1-methyl-4-piperidinemethanol,1-methylpiperidin-4-yl methanol,n-methyl-4-piperidinemethanol,4-piperidinemethanol, 1-methyl,1-methyl-4-piperidinyl methanol,4-hydroxymethyl-1-methylpiperidine,1-methylpiperidine-4-methanol,n-methyl-4-hydroxymethylpiperidine,1-methyl-piperidin-4-yl-methanol,1-methyl-4-piperidyl methan-1-ol
271971
(1-methylpiperidin-4-yl)methanol
CN1CCC(CC1)CO
20691-89-8
C7H15NO
129.2
KJZLJGZZDNGGCA-UHFFFAOYSA-N
1-methyl-4-piperidinemethanol,1-methylpiperidin-4-yl methanol,n-methyl-4-piperidinemethanol,4-piperidinemethanol, 1-methyl,1-methyl-4-piperidinyl methanol,4-hydroxymethyl-1-methylpiperidine,1-methylpiperidine-4-methanol,n-methyl-4-hydroxymethylpiperidine,1-methyl-piperidin-4-yl-methanol,1-methyl-4-piperidyl methan-1-ol
271971
(1-methylpiperidin-4-yl)methanol
CN1CCC(CC1)CO
Specific Gravity 0.98
Quantity 1g
Density 0.9800g/mL
Infrared Spectrum Authentic
Boiling Point 200.0°C
Color Colorless to Yellow
Packaging Glass bottle
Melting Point 29.0°C to 31.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 129.2
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1-Methyl-4-piperidinemethanol
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning