Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-5-phenyl-1H-pyrazole-3-carboxylic acid, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
10199-53-8
C11H10N2O2
202.21
ZDUDGEACIDDEPL-UHFFFAOYSA-N
1-methyl-5-phenyl-1h-pyrazole-3-carboxylic acid,1h-pyrazole-3-carboxylicacid, 1-methyl-5-phenyl,1h-pyrazole-3-carboxylic acid, 1-methyl-5-phenyl,3-carboxy-1-methyl-5-phenyl-1h-pyrazole,1-methyl-5-phenyl pyrazole-3-carboxylic acid,3-carboxy-1-methyl-1h-pyrazol-5-yl benzene,1-methyl-5-phenyl-1h-3-pyrazolecarboxylic acid
7537619
1-methyl-5-phenylpyrazole-3-carboxylic acid
CN1C(=CC(=N1)C(=O)O)C2=CC=CC=C2
10199-53-8
C11H10N2O2
202.21
ZDUDGEACIDDEPL-UHFFFAOYSA-N
1-methyl-5-phenyl-1h-pyrazole-3-carboxylic acid,1h-pyrazole-3-carboxylicacid, 1-methyl-5-phenyl,1h-pyrazole-3-carboxylic acid, 1-methyl-5-phenyl,3-carboxy-1-methyl-5-phenyl-1h-pyrazole,1-methyl-5-phenyl pyrazole-3-carboxylic acid,3-carboxy-1-methyl-1h-pyrazol-5-yl benzene,1-methyl-5-phenyl-1h-3-pyrazolecarboxylic acid
7537619
1-methyl-5-phenylpyrazole-3-carboxylic acid
CN1C(=CC(=N1)C(=O)O)C2=CC=CC=C2
Quantity 5g
Infrared Spectrum Authentic
Melting Point 144.0°C to 145.0°C
Assay Percent Range 96% min. (HPLC)
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 1-Methyl-5-phenyl-1H-pyrazole-3-carboxylic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning