Alfa Aesar™ 1-Methyl-5-(tri-n-butylstannyl)imidazole, 90+%

Manufacturer: Alfa Aesar™ H51405MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51405MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 147716-03-8
    Molecular Formula C16H32N2Sn
    Formula Weight 371.15
    MDL Number MFCD01319031
    UN Number UN2788
    Synonym 5-(Tri-n-butylstannyl)-1-methylimidazole
    Chemical Name or Material 1-Methyl-5-(tri-n-butylstannyl)imidazole

    Specifications

    Formula Weight 371.15
    UN Number UN2788
    Synonym 5-(Tri-n-butylstannyl)-1-methylimidazole
    Quantity 250mg
    Health Hazard 2 GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P260-P280-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C16H32N2Sn
    MDL Number MFCD01319031
    CAS 147716-03-8
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    Chemical Name or Material 1-Methyl-5-(tri-n-butylstannyl)imidazole
    Assay ≥90%

    Description

    Applications
    1-Methyl-5-(tri-n-butylstannyl)imidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

    Solubility
    Sparingly soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P280-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit